Search
Search

gcy-19| Gene Symbol: gcy-19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001361852.3 | NP_001348701.1 | 20 | 3203 | 115 | |
| NM_001361853.3 | NP_001348700.1 | 20 - 21 | 3273 | 115 | ||
| NM_001361854.3 | NP_001348702.1 | 21 | 3236 | 115 | ||
Set Membership: |
|