Search
Search

clpp-1| Gene Symbol: clpp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267310.4 | NP_001254239.1 | 4 - 5 | 574 | 64 | |
| NM_001267311.3 | NP_001254240.1 | 4 - 5 | 523 | 64 | ||
| NM_001267312.3 | NP_001254241.1 | 2 - 3 | 247 | 64 | ||
| NR_051588.1 | 1 - 2 | 103 | 64 | |||
Set Membership: |
|