Search
Search

smg-6| Gene Symbol: smg-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381779.2 | NP_001367726.1 | 1 - 2 | 376 | 76 | |
| NM_065164.8 | NP_497565.1 | 1 - 2 | 391 | 76 | ||
| NM_065165.8 | NP_497566.3 | 1 - 2 | 377 | 76 | ||
Set Membership: |
|