Search
Search

| Gene Symbol: ulp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065694.8 | NP_498095.3 | 4 - 5 | 641 | 75 | |
| GenBank mRNA | AB095020.1 | - | 4 - 5 | 581 | 75 | |
| Gene Symbol: T10F2.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001382862.1 | 4 - 5 | 602 | 75 | ||
| Gene Symbol: prp-19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001382863.1 | 7 - 8 | 2684 | 75 | ||
Set Membership: |
|