Search
Search

cla-1| Gene Symbol: cla-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001307214.3 | NP_001294143.1 | 18 | 6698 | 68 | |
| NM_001307220.4 | NP_001294149.2 | 31 | 25538 | 68 | ||
| NM_001380338.1 | NP_001367468.1 | 1 | 2465 | 68 | ||
Set Membership: |
|