Search
Search

unc-43| Gene Symbol: unc-43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028127.6 | NP_001023298.1 | 3 - 4 | 365 | 79 | |
| NM_001268551.3 | NP_001255480.1 | 4 - 5 | 384 | 79 | ||
| NM_001372918.2 | NP_001359633.1 | 4 | 445 | 79 | ||
| NM_001380439.2 | NP_001366993.1 | 4 - 5 | 505 | 79 | ||
| NM_001380440.2 | NP_001366994.1 | 4 - 5 | 459 | 79 | ||
| NM_001380441.2 | NP_001366996.1 | 1 - 2 | 208 | 79 | ||
| NM_001380442.1 | NP_001366992.1 | 2 - 3 | 289 | 79 | ||
| NM_001380443.1 | NP_001366997.1 | 1 - 2 | 54 | 79 | ||
| NM_001383186.1 | 3 - 4 | 365 | 79 | |||
| NM_001383187.1 | 3 - 4 | 359 | 79 | |||
| GenBank mRNA | AF180735.1 | - | 2 - 3 | 267 | 79 | |
| AF233262.1 | - | 1 - 2 | 54 | 79 | ||
| AF233263.1 | - | 1 - 2 | 54 | 79 | ||
| AF233264.1 | - | 1 - 2 | 54 | 79 | ||
| AF233265.1 | - | 1 - 2 | 54 | 79 | ||
| AF233266.1 | - | 1 - 2 | 54 | 79 | ||
| AF233267.1 | - | 1 - 2 | 54 | 79 | ||
| AF255956.1 | - | 1 - 2 | 54 | 79 | ||
Set Membership: |
|