Search
Search

lec-11| Gene Symbol: lec-11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028020.5 | NP_001023191.1 | 2 - 3 | 283 | 146 | |
| NR_131549.1 | 2 - 3 | 112 | 146 | |||
| GenBank mRNA | AB038502.2 | - | 2 - 3 | 208 | 146 | |
Set Membership: |
|