Search
Search

flp-34| Gene Symbol: flp-34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383273.1 | 11 - 12 | 3619 | 60 | ||
| NM_070964.6 | NP_503365.1 | 3 - 4 | 304 | 60 | ||
| NR_132496.1 | 1 - 2 | 63 | 60 | |||
Set Membership: |
|