Search
Search

acl-6| Gene Symbol: acl-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028598.4 | NP_001023769.1 | 10 - 11 | 1822 | 68 | |
| NM_001028599.5 | NP_001023770.1 | 10 - 11 | 1824 | 68 | ||
Set Membership: |
|