Search
Search

twk-13| Gene Symbol: twk-13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129512.4 | NP_001122984.1 | 5 - 6 | 994 | 108 | |
| NM_001276939.4 | NP_001263868.1 | 7 - 8 | 749 | 108 | ||
| NM_001276940.3 | NP_001263869.1 | 6 - 7 | 592 | 108 | ||
| NM_073690.10 | NP_506091.2 | 5 - 6 | 523 | 108 | ||
Set Membership: |
|