Search
Search
| Gene Symbol: XKR9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014109283.2 | XP_013964758.1 | 5 | 867 | 123 | |
| XM_014109286.2 | XP_013964761.1 | 4 | 782 | 123 | ||
| XM_022412238.1 | XP_022267946.1 | 7 | 1149 | 123 | ||
| XM_022412239.1 | XP_022267947.1 | 6 | 1139 | 123 | ||
| XM_022412240.1 | XP_022267948.1 | 6 | 964 | 123 | ||
Set Membership: |
|