Search
Search
| Gene Symbol: HJV | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005630732.2 | XP_005630789.1 | 4 | 628 | 153 | |
| XM_005630733.3 | XP_005630790.1 | 5 | 802 | 153 | ||
| XM_005630734.3 | XP_005630791.1 | 3 | 380 | 153 | ||
| XM_014120583.2 | XP_013976058.1 | 4 | 1424 | 153 | ||
| XM_022404943.1 | XP_022260651.1 | 4 | 677 | 153 | ||
| XM_022404944.1 | XP_022260652.1 | 3 | 579 | 153 | ||
| XM_022404945.1 | XP_022260653.1 | 2 | 542 | 153 | ||
| XM_022404946.1 | XP_022260654.1 | 2 | 492 | 153 | ||
Set Membership: |
|