Search
Search

Ifngr2| Gene Symbol: Ifngr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007642060.1 | XP_007640250.1 | 4 - 5 | 831 | 71 | |
| XM_007642065.1 | XP_007640255.1 | 4 - 5 | 862 | 71 | ||
| GenBank mRNA | JI865507.1 | - | 3 - 4 | 449 | 71 | |
Set Membership: |
|