Search
Search

Nkiras2| Gene Symbol: Nkiras2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504639.2 | XP_003504687.1 | 2 - 3 | 248 | 68 | |
| XM_007645133.1 | XP_007643323.1 | 2 - 3 | 135 | 68 | ||
Set Membership: |
|