Search
Search

Prss27| Gene Symbol: Prss27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007650146.1 | XP_007648336.1 | 3 - 4 | 397 | 76 | |
| XM_007650147.1 | XP_007648337.1 | 3 - 4 | 588 | 76 | ||
| GenBank mRNA | JI865189.1 | - | 2 - 3 | 146 | 76 | |
| JP049856.1 | - | 2 - 3 | 145 | 76 | ||
Set Membership: |
|