Search
Search
| Gene Symbol: SNRK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018066940.1 | XP_017922429.1 | 5 - 6 | 1221 | 62 | |
| XM_018066941.1 | XP_017922430.1 | 4 - 5 | 1014 | 62 | ||
| XM_018066942.1 | XP_017922431.1 | 5 - 6 | 1064 | 62 | ||
| XM_018066943.1 | XP_017922432.1 | 3 - 4 | 952 | 62 | ||
| XM_018066944.1 | XP_017922433.1 | 6 - 7 | 1126 | 62 | ||
| GenBank mRNA | JO405679.2 | - | 2 - 3 | 726 | 62 | |
| JT762829.1 | - | 2 - 3 | 258 | 62 | ||
Set Membership: |
|