Search
Search
| Gene Symbol: THEM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005677585.3 | XP_005677642.1 | 1 - 2 | 231 | 70 | |
| GenBank mRNA | JO437566.2 | - | 5 - 6 | 1035 | 70 | |
| JT669341.1 | - | 1 - 2 | 202 | 70 | ||
Set Membership: |
|