Search
Search

otp| Gene Symbol: otp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043098.2 | NP_001036563.1 | 3 - 4 | 838 | 54 | |
| NM_001103918.3 | NP_001097388.2 | 4 - 5 | 1326 | 54 | ||
| NM_001299721.1 | NP_001286650.1 | 3 - 4 | 838 | 54 | ||
| NM_001299722.1 | NP_001286651.1 | 3 - 4 | 817 | 54 | ||
| NM_001299723.1 | NP_001286652.1 | 4 - 5 | 1326 | 54 | ||
| NM_001299724.1 | NP_001286653.1 | 4 - 5 | 1347 | 54 | ||
| NM_001299725.1 | NP_001286654.1 | 4 - 5 | 1347 | 54 | ||
| NM_001369956.1 | NP_001356966.1 | 4 - 5 | 1326 | 54 | ||
| NM_079075.5 | NP_523799.3 | 3 - 4 | 817 | 54 | ||
| GenBank mRNA | BT023887.1 | - | 3 - 4 | 826 | 54 | |
| BT030422.1 | - | 4 - 5 | 1307 | 54 | ||
Set Membership: |
|