Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Dm01803616_g1
          See other PVR GE Assays ›
          Gene Symbol:
          Pvr
          Gene Name:
          PDGF- and VEGF-receptor related
          Gene Aliases:
          8222, CG8222, CT24332, DmVEGFR, Dmel\CG8222, Dmel_CG8222, PVR, PvR, VEGFR, VEGFR-A, VGR1, Vegf, Vegfr, Vegfr-b, Vegfr-c, Vgr1, pvr, stai, vgr1
          Chromosome Location:
          Chr.2L: 8220980 - 8239878 on Build Release_6_plus_ISO1_MT
          Species:
          Drosophila melanogaster
          Species Specific ID (Flybase ID):
          FBgn0032006
          Assay ID Dm01803616_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pvr
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001169438.2 NP_001162909.1 10 - 11 3293 58
          NM_001169439.2 NP_001162910.1 11 - 12 2880 58
          NM_001169440.2 NP_001162911.1 11 - 12 2880 58
          NM_001201791.2 NP_001188720.1 9 - 10 2649 58
          NM_001201792.2 NP_001188721.1 10 - 11 2549 58
          NM_001273305.1 NP_001260234.1 10 - 11 2853 58
          NM_001273306.1 NP_001260235.1 9 - 10 2646 58
          NM_078785.4 NP_523509.2 10 - 11 2676 58
          NM_164801.3 NP_723365.1 10 - 11 2676 58
          NM_175983.3 NP_787997.2 10 - 11 2658 58
          NM_205925.3 NP_995647.2 9 - 10 2646 58
          NM_205926.3 NP_995648.1 10 - 11 2711 58
          NM_205927.3 NP_995649.1 10 - 11 3293 58
          GenBank mRNA BT050546.1 - 9 - 10 2425 58
          BT133154.1 - 11 - 12 2849 58

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 Drosophila melanogaster

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          response to tumor cell
          negative regulation of humoral immune response
          transmembrane receptor protein tyrosine kinase signaling pathway
          multicellular organism development
          border follicle cell migration
          dorsal closure
          ventral cord development
          salivary gland morphogenesis
          Malpighian tubule morphogenesis
          glial cell migration
          positive regulation of phosphatidylinositol 3-kinase signaling
          cell projection assembly
          hemopoiesis
          positive regulation of cell migration
          positive regulation of kinase activity
          hemocyte migration
          larval lymph gland hemopoiesis
          wound healing, spreading of epidermal cells
          intestinal stem cell homeostasis
          negative regulation of apoptotic process
          innate immune response
          positive regulation of cell size
          positive regulation of JNK cascade
          imaginal disc fusion, thorax closure
          vascular endothelial growth factor receptor signaling pathway
          lymph gland development
          imaginal disc-derived male genitalia morphogenesis
          positive regulation of protein kinase B signaling
          anterior Malpighian tubule development
          positive regulation of ERK1 and ERK2 cascade
          positive regulation of border follicle cell migration
          positive regulation of TORC1 signaling
          transmembrane receptor protein tyrosine kinase activity
          protein binding
          ATP binding
          vascular endothelial growth factor binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline