Search
Search

Gli| Gene Symbol: Gli | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057254.4 | NP_476602.1 | 5 - 6 | 2145 | 96 | |
| NM_165127.2 | NP_723928.1 | 6 - 7 | 2236 | 96 | ||
| NM_165128.2 | NP_723929.1 | 5 - 6 | 2025 | 96 | ||
| NM_165129.2 | NP_723930.1 | 5 - 6 | 2057 | 96 | ||
| NM_165130.2 | NP_723931.2 | 5 - 6 | 2518 | 96 | ||
| GenBank mRNA | BT133203.1 | - | 5 - 6 | 2142 | 96 | |
Set Membership: |
|