Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Dm01820466_g1
See other ZIP GE Assays ›
Gene Symbol:
zip
Gene Name:
zipper
Gene Aliases:
CG15792, DROMHC, Dm nmII, Dmel\CG15792, Dmel_CG15792, DmnmII, Dronm-MII, E(br), MHC, Mhc-c, Mhc-c[1], Myo, Myo II, Myo-II, Myo-II HC, Myo15, Myo2, MyoII, NMM, NMMII, ZIP, Zip, anon-WO0140519.37, l(2)02957, l(2)17F1, myo II, myoII, myosin-II, nmMHC, nmy-2, zip/MyoII, zpr
Chromosome Location:
Chr.2R: 24990570 - 25011965 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0265434
Assay ID
Dm01820466_g1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2,900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2,900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2,900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: zip
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_079136.4
NP_523860.2
6 - 7
906
127
NM_166704.4
NP_726506.1
6 - 7
842
127
Back To Top
More Information
Set Membership:
Amplicon length greater than or equal to 101
Drosophila melanogaster
Gene Ontology Categories:
Function(s)
Process(es)
mitotic cytokinesis
muscle contraction
border follicle cell migration
dorsal closure
dorsal closure, spreading of leading edge cells
salivary gland morphogenesis
Malpighian tubule morphogenesis
eye-antennal disc morphogenesis
anterior midgut development
head involution
muscle attachment
myofibril assembly
myosin II filament assembly
maintenance of protein location in cell
cuticle pattern formation
regulation of tube length, open tracheal system
spiracle morphogenesis, open tracheal system
imaginal disc-derived wing hair organization
wound healing
sarcomere organization
dorsal closure, leading edge cell differentiation
dorsal closure, amnioserosa morphology change
compartment boundary maintenance
morphogenesis of an epithelial fold
left/right axis specification
cellular response to mechanical stimulus
regulation of establishment of planar polarity
regulation of myoblast fusion
positive regulation of border follicle cell migration
motor activity
protein binding
ATP binding
myosin light chain binding
actin filament binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline