Search
Search

CaMKII| Gene Symbol: CaMKII | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014696.2 | NP_001014696.1 | 2 - 3 | 362 | 98 | |
| NM_001169360.2 | NP_001162831.1 | 2 - 3 | 841 | 98 | ||
| NM_001169361.2 | NP_001162832.1 | 3 - 4 | 258 | 98 | ||
| NM_001169362.2 | NP_001162833.1 | 2 - 3 | 192 | 98 | ||
| NM_001272153.1 | NP_001259082.1 | 2 - 3 | 362 | 98 | ||
| NM_001272154.1 | NP_001259083.1 | 2 - 3 | 362 | 98 | ||
| NM_001272155.1 | NP_001259084.1 | 2 - 3 | 362 | 98 | ||
| NM_079896.4 | NP_524635.3 | 2 - 3 | 362 | 98 | ||
| NM_166810.5 | NP_726633.2 | 2 - 3 | 192 | 98 | ||
| NM_166811.2 | NP_726634.1 | 2 - 3 | 276 | 98 | ||
| NM_166812.3 | NP_726635.2 | 2 - 3 | 362 | 98 | ||
| NM_166813.3 | NP_726636.2 | 2 - 3 | 362 | 98 | ||
| GenBank mRNA | BT050453.1 | - | 2 - 3 | 780 | 98 | |
| M74583.1 | - | 2 - 3 | 316 | 98 | ||
Set Membership: |
|