Search
Search

mRpS6| Gene Symbol: mRpS6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259694.2 | NP_001246623.1 | 1 - 2 | 201 | 55 | |
| NM_079201.5 | NP_523925.1 | 2 - 3 | 108 | 55 | ||
| GenBank mRNA | AY070674.1 | - | 1 - 2 | 196 | 55 | |
Set Membership: |
|