Search
Search

Tak1| Gene Symbol: Tak1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079356.3 | NP_524080.1 | 6 - 7 | 2293 | 73 | |
| GenBank mRNA | AF199466.1 | - | 6 - 7 | 2247 | 73 | |
| AY051953.1 | - | 6 - 7 | 2266 | 73 | ||
Set Membership: |
|