Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Dm01841974_g1
See other N GE Assays ›
Gene Symbol:
N
Gene Name:
Notch
Gene Aliases:
1.1, 16-178, 16-55, Ax, CG3936, CT13012, Chp, Co, Dmel\CG3936, Dmel_CG3936, EG:140G11.1, EG:163A10.2, NECD, NICD, Nicd, RICN, anon-EST:Liang-1.12, clone 1.12, co, dNotch, fa, l(1)3Cb, l(1)Ax, l(1)N, n[fah], nd, notch, shd, spl, swb
Chromosome Location:
Chr.X: 3134870 - 3172221 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0004647
Assay ID
Dm01841974_g1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 250 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2,900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2,900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2,900 rxns
Availability
Inventoried
Catalog #
4331182
Price
284.00
Your Price
Online offer:
266.65
266.65
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: N
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001258581.2
NP_001245510.1
8 - 9
8569
67
NM_057511.4
NP_476859.2
8 - 9
8569
67
Back To Top
More Information
Set Membership:
Amplicon length less than or equal to 70
3' Most
Drosophila melanogaster
Inventoried
Panther Classification:
Molecular Function -
cell adhesion molecule
Gene Ontology Categories:
Function(s)
Process(es)
compound eye morphogenesis
positive regulation of neuroblast proliferation
defense response to insect
regulation of glycolytic process
actin filament organization
Notch signaling pathway
germarium-derived egg chamber formation
ovarian follicle cell migration
border follicle cell migration
regulation of mitotic cell cycle
dorsal closure
neuroblast fate determination
glial cell fate determination
neuroblast proliferation
axon guidance
peripheral nervous system development
sensory organ development
foregut morphogenesis
eye-antennal disc morphogenesis
imaginal disc-derived wing vein specification
imaginal disc-derived wing morphogenesis
leg disc morphogenesis
imaginal disc-derived leg morphogenesis
mesoderm development
muscle cell fate determination
anesthesia-resistant memory
long-term memory
motor neuron axon guidance
positive regulation of cell proliferation
determination of adult lifespan
glial cell migration
asymmetric cell division
chaeta morphogenesis
imaginal disc-derived wing margin morphogenesis
response to symbiont
dorsal/ventral axis specification
glial cell differentiation
positive regulation of gene expression
negative regulation of gene expression
neuroblast development
second mitotic wave involved in compound eye morphogenesis
imaginal disc-derived leg joint morphogenesis
sensory organ precursor cell fate determination
chaeta development
cell differentiation
ovarian follicle cell development
ovarian follicle cell stalk formation
germ-line stem cell population maintenance
oocyte localization involved in germarium-derived egg chamber formation
epithelial cell type specification, open tracheal system
embryonic hemopoiesis
larval lymph gland hemopoiesis
lamellocyte differentiation
hemocyte proliferation
negative regulation of lamellocyte differentiation
eye-antennal disc development
wing disc pattern formation
imaginal disc-derived leg segmentation
female germ-line stem cell population maintenance
intestinal stem cell homeostasis
regulation of growth
regulation of cardioblast cell fate specification
crystal cell differentiation
positive regulation of crystal cell differentiation
positive regulation of neuron apoptotic process
cell dedifferentiation
cell fate commitment
negative regulation of compound eye photoreceptor development
R7 cell differentiation
regulation of cell differentiation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
lateral inhibition
retinal cell programmed cell death
compound eye retinal cell programmed cell death
muscle cell cellular homeostasis
dorsal appendage formation
R1/R6 cell differentiation
R3/R4 cell differentiation
wing disc dorsal/ventral pattern formation
oogenesis
lymph gland development
myoblast development
neuron fate determination
neuron fate specification
compound eye development
imaginal disc-derived male genitalia morphogenesis
stem cell differentiation
regulation of neurogenesis
negative regulation of neurogenesis
neurological system process
regulation of filopodium assembly
germ-line stem-cell niche homeostasis
formation of a compartment boundary
compartment boundary maintenance
morphogenesis of an epithelial fold
epithelial cell proliferation involved in Malpighian tubule morphogenesis
Malpighian tubule tip cell differentiation
neuronal stem cell population maintenance
positive regulation of G1/S transition of mitotic cell cycle
regulation of neuroblast proliferation
regulation of stem cell division
negative regulation of cell-cell adhesion mediated by cadherin
chromatin binding
transmembrane signaling receptor activity
calcium ion binding
protein binding
histone acetyltransferase binding
WW domain binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline