Search
Search

kn| Gene Symbol: kn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103847.2 | NP_001097317.1 | 9 - 10 | 1544 | 63 | |
| NM_080074.4 | NP_524813.2 | 8 - 9 | 1665 | 63 | ||
| NM_166070.3 | NP_725419.2 | 8 - 9 | 1665 | 63 | ||
| GenBank mRNA | BT150395.1 | - | 8 - 9 | 1663 | 63 | |
Set Membership: |
|