Search
Search

Sap47| Gene Symbol: Sap47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275688.1 | NP_001262617.1 | 6 - 7 | 1544 | 75 | |
| NM_001300421.1 | NP_001287350.1 | 5 - 6 | 1940 | 75 | ||
| NM_001370053.1 | NP_001356917.1 | 5 - 6 | 1940 | 75 | ||
| NM_001370054.1 | NP_001356918.1 | 6 - 7 | 1574 | 75 | ||
| NM_079647.4 | NP_524371.1 | 5 - 6 | 1921 | 75 | ||
| NM_169684.3 | NP_732088.1 | 6 - 7 | 1990 | 75 | ||
| NM_169685.3 | NP_732089.1 | 7 - 8 | 1598 | 75 | ||
| NM_169686.3 | NP_732090.1 | 7 - 8 | 1926 | 75 | ||
| NM_169687.3 | NP_732091.1 | 7 - 8 | 1744 | 75 | ||
| NM_169688.3 | NP_732092.2 | 5 - 6 | 1940 | 75 | ||
| NM_169689.3 | NP_732093.1 | 6 - 7 | 1574 | 75 | ||
| NM_169690.3 | NP_732094.1 | 6 - 7 | 1990 | 75 | ||
| GenBank mRNA | AY051871.1 | - | 6 - 7 | 1938 | 75 | |
| BT126317.1 | - | 6 - 7 | 1177 | 75 | ||
| X80110.1 | - | 1212 | 75 | |||
| X80111.1 | - | 1540 | 75 | |||
Set Membership: |
|