Search
Search

Bili| Gene Symbol: Bili | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170251.2 | NP_001163722.1 | 4 - 5 | 1334 | 73 | |
| NM_001170252.2 | NP_001163723.1 | 4 - 5 | 1323 | 73 | ||
| NM_001170253.2 | NP_001163724.1 | 4 - 5 | 1418 | 73 | ||
| NM_001170254.2 | NP_001163725.1 | 4 - 5 | 1354 | 73 | ||
| NM_001170255.1 | NP_001163726.1 | 4 - 5 | 1474 | 73 | ||
| NM_001300592.1 | NP_001287521.1 | 4 - 5 | 1727 | 73 | ||
| NM_143085.3 | NP_651342.3 | 4 - 5 | 1431 | 73 | ||
| GenBank mRNA | AY094737.1 | - | 4 - 5 | 1393 | 73 | |
Set Membership: |
|