Search
Search

RhoGAP1A| Gene Symbol: RhoGAP1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103385.2 | NP_001096855.1 | 2 - 3 | 636 | 72 | |
| NM_001103386.2 | NP_001096856.1 | 2 - 3 | 636 | 72 | ||
| NM_001297795.1 | NP_001284724.1 | 2 - 3 | 640 | 72 | ||
| NM_001297796.1 | NP_001284725.1 | 2 - 3 | 640 | 72 | ||
| GenBank mRNA | BT024449.1 | - | 2 - 3 | 598 | 72 | |
Set Membership: |
|