Search
Search

anxa2a| Gene Symbol: anxa2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181761.2 | NP_861426.1 | 10 - 11 | 822 | 90 | |
| GenBank mRNA | BC056699.1 | - | 10 - 11 | 822 | 90 | |
| BC062531.1 | - | 10 - 11 | 814 | 90 | ||
Set Membership: |
|