Search
Search

fgf14| Gene Symbol: fgf14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012382.1 | NP_001012382.1 | 1 - 2 | 192 | 78 | |
| GenBank mRNA | AB195620.1 | - | 1 - 2 | 192 | 78 | |
| BC162852.1 | - | 1 - 2 | 258 | 78 | ||
| BC162900.1 | - | 1 - 2 | 258 | 78 | ||
Set Membership: |
|