Search
Search

ephb3a| Gene Symbol: ephb3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131097.2 | NP_571172.2 | 3 - 4 | 847 | 91 | |
| XM_009297449.3 | XP_009295724.1 | 3 - 4 | 847 | 91 | ||
| XM_017357616.2 | XP_017213105.1 | 3 - 4 | 847 | 91 | ||
| GenBank mRNA | AF369382.1 | - | 3 - 4 | 414 | 91 | |
| KM655700.1 | - | 3 - 4 | 414 | 91 | ||
Set Membership: |
|