Search
Search

nipa1| Gene Symbol: nipa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006062.1 | NP_001006062.1 | 4 - 5 | 569 | 119 | |
| GenBank mRNA | BC083262.1 | - | 4 - 5 | 569 | 119 | |
| BC164079.1 | - | 4 - 5 | 448 | 119 | ||
Set Membership: |
|