Search
Search

sp5l| Gene Symbol: sp5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194371.2 | NP_919352.1 | 1 - 2 | 385 | 69 | |
| GenBank mRNA | AB097827.1 | - | 1 - 2 | 60 | 69 | |
| AY338748.1 | - | 1 - 2 | 385 | 69 | ||
| BC094980.1 | - | 1 - 2 | 477 | 69 | ||
| BC116567.1 | - | 1 - 2 | 406 | 69 | ||
Set Membership: |
|