Search
Search

brk1| Gene Symbol: brk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002097.1 | NP_001002097.1 | 2 - 3 | 225 | 65 | |
| XM_005172903.4 | XP_005172960.1 | 4 - 5 | 694 | 65 | ||
| GenBank mRNA | BC046048.1 | - | 2 - 3 | 244 | 65 | |
| BC071524.1 | - | 2 - 3 | 225 | 65 | ||
| BC154586.1 | - | 4 - 5 | 650 | 65 | ||
Set Membership: |
|