Search
Search

nup35| Gene Symbol: nup35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200066.1 | NP_956360.1 | 6 - 7 | 613 | 120 | |
| XM_009304508.3 | XP_009302783.1 | 6 - 7 | 722 | 120 | ||
| GenBank mRNA | BC061958.1 | - | 6 - 7 | 613 | 120 | |
Set Membership: |
|