Search
Search

mtmr1b| Gene Symbol: mtmr1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014330.2 | NP_001014352.1 | 3 - 4 | 492 | 66 | |
| GenBank mRNA | BC091956.1 | - | 3 - 4 | 496 | 66 | |
| BC165859.1 | - | 3 - 4 | 259 | 66 | ||
Set Membership: |
|