Search
Search

arhgap4a| Gene Symbol: arhgap4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200444.1 | NP_956738.1 | 6 - 7 | 641 | 107 | |
| XM_005162155.4 | XP_005162212.1 | 6 - 7 | 741 | 107 | ||
| GenBank mRNA | BC054931.1 | - | 6 - 7 | 641 | 107 | |
Set Membership: |
|