Search
Search

lgals3b| Gene Symbol: lgals3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214693.2 | NP_999858.2 | 3 - 4 | 331 | 112 | |
| GenBank mRNA | AY421705.1 | - | 3 - 4 | 320 | 112 | |
| BC093113.1 | - | 3 - 4 | 341 | 112 | ||
| BC107990.1 | - | 3 - 4 | 331 | 112 | ||
Set Membership: |
|