Search
Search

gnrh2| Gene Symbol: gnrh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181439.4 | NP_852104.3 | 2 - 3 | 308 | 101 | |
| GenBank mRNA | AF511531.1 | - | 2 - 3 | 308 | 101 | |
| AY094357.1 | - | 1 - 2 | 144 | 101 | ||
| AY657018.1 | - | 2 - 3 | 308 | 101 | ||
| BC162945.1 | - | 2 - 3 | 261 | 101 | ||
| BC162951.1 | - | 2 - 3 | 261 | 101 | ||
Set Membership: |
|