Search
Search

anxa1b| Gene Symbol: anxa1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181759.1 | NP_861424.1 | 5 - 6 | 493 | 117 | |
| GenBank mRNA | AY178794.1 | - | 5 - 6 | 493 | 117 | |
| BC092685.1 | - | 6 - 7 | 513 | 117 | ||
| BC164147.1 | - | 5 - 6 | 496 | 117 | ||
Set Membership: |
|