Search
Search

eif4bb| Gene Symbol: eif4bb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214718.1 | NP_999883.1 | 2 - 3 | 172 | 69 | |
| XM_005166014.4 | XP_005166071.1 | 2 - 3 | 194 | 69 | ||
| XM_021476791.1 | XP_021332466.1 | 2 - 3 | 193 | 69 | ||
| GenBank mRNA | AY576997.1 | - | 2 - 3 | 172 | 69 | |
| BC066675.1 | - | 1 - 2 | 149 | 69 | ||
Set Membership: |
|