Search
Search

dnaaf4| Gene Symbol: dnaaf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205688.1 | NP_991251.1 | 3 - 4 | 422 | 72 | |
| GenBank mRNA | AY428510.1 | - | 3 - 4 | 518 | 72 | |
| BC065881.1 | - | 3 - 4 | 422 | 72 | ||
| BC165842.1 | - | 3 - 4 | 438 | 72 | ||
Set Membership: |
|