Search
Search

oxct1b| Gene Symbol: oxct1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077150.1 | NP_001070618.1 | 2 - 3 | 259 | 122 | |
| GenBank mRNA | BC124298.1 | - | 2 - 3 | 259 | 122 | |
| BC154254.1 | - | 2 - 3 | 241 | 122 | ||
| BC155216.1 | - | 2 - 3 | 281 | 122 | ||
| BC164688.1 | - | 2 - 3 | 243 | 122 | ||
Set Membership: |
|