Search
Search

CCNT1| Gene Symbol: CCNT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081846.1 | NP_001075315.1 | 5 - 6 | 499 | 70 | |
| GenBank mRNA | AF137509.1 | - | 5 - 6 | 499 | 70 | |
| AF190905.1 | - | 5 - 6 | 499 | 70 | ||
Set Membership: |
|