Search
Search

PPP6R3| Gene Symbol: PPP6R3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113235.1 | NP_001106706.1 | 9 - 10 | 1221 | 64 | |
| XM_023653807.1 | XP_023509575.1 | 1253 | 64 | |||
| XM_023653808.1 | XP_023509576.1 | 1077 | 64 | |||
| XM_023653810.1 | XP_023509578.1 | 1085 | 64 | |||
| XM_023653811.1 | XP_023509579.1 | 1253 | 64 | |||
| XM_023653812.1 | XP_023509580.1 | 1253 | 64 | |||
| XM_023653813.1 | XP_023509581.1 | 1253 | 64 | |||
| XM_023653814.1 | XP_023509582.1 | 1253 | 64 | |||
| XM_023653815.1 | XP_023509583.1 | 1253 | 64 | |||
| XM_023653816.1 | XP_023509584.1 | 1253 | 64 | |||
| XM_023653817.1 | XP_023509585.1 | 1253 | 64 | |||
| XM_023653818.1 | XP_023509586.1 | 1253 | 64 | |||
| XM_023653819.1 | XP_023509587.1 | 1253 | 64 | |||
| XM_023653821.1 | XP_023509589.1 | 1253 | 64 | |||
| XM_023653822.1 | XP_023509590.1 | 774 | 64 | |||
| XM_023653823.1 | XP_023509591.1 | 833 | 64 | |||
| XM_023653824.1 | XP_023509592.1 | 1253 | 64 | |||
| GenBank mRNA | AB371895.1 | - | 9 - 10 | 1221 | 64 | |
| AB371896.1 | - | 9 - 10 | 1221 | 64 | ||
| AB371897.1 | - | 9 - 10 | 1221 | 64 | ||
Set Membership: |
|