Search
Search

UBE2D3| Gene Symbol: UBE2D3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031153.1 | NP_001026324.1 | 4 - 5 | 273 | 147 | |
| XM_015285429.1 | XP_015140915.1 | 4 - 5 | 412 | 147 | ||
| XM_015285430.1 | XP_015140916.1 | 4 - 5 | 412 | 147 | ||
| XM_015285431.2 | XP_015140917.1 | 4 - 5 | 310 | 147 | ||
| XM_025149681.1 | XP_025005449.1 | 2 - 3 | 103 | 147 | ||
| GenBank mRNA | AJ719875.1 | - | 4 - 5 | 273 | 147 | |
Set Membership: |
|