Search
Search

TPI1| Gene Symbol: TPI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205451.1 | NP_990782.1 | 6 - 7 | 740 | 150 | |
| GenBank mRNA | EF117328.1 | - | 2 - 3 | 157 | 150 | |
| M11314.1 | - | 6 - 7 | 740 | 150 | ||
Set Membership: |
|