Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00172783_m1
          See other EZH2 GE Assays ›
          Gene Symbol:
          EZH2
          Gene Name:
          enhancer of zeste 2 polycomb repressive complex 2 subunit
          Gene Aliases:
          ENX-1, ENX1, EZH1, EZH2b, KMT6, KMT6A, WVS, WVS2
          Chromosome Location:
          Chr.7: 148807372 - 148884662 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs00172783_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: EZH2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001203247.1 NP_001190176.1 4 - 5 558 137
          NM_001203248.1 NP_001190177.1 4 - 5 531 137
          NM_001203249.1 NP_001190178.1 4 - 5 658 137
          NM_004456.4 NP_004447.2 4 - 5 558 137
          XM_005249962.4 XP_005250019.1 4 - 5 2999 137
          XM_005249963.4 XP_005250020.1 4 - 5 2972 137
          XM_005249964.4 XP_005250021.1 4 - 5 2973 137
          XM_011515883.2 XP_011514185.1 5 - 6 533 137
          XM_011515884.2 XP_011514186.1 4 - 5 665 137
          XM_011515885.2 XP_011514187.1 4 - 5 2974 137
          XM_011515886.2 XP_011514188.1 5 - 6 682 137
          XM_011515887.2 XP_011514189.1 4 - 5 531 137
          XM_011515888.2 XP_011514190.1 4 - 5 653 137
          XM_011515889.2 XP_011514191.1 4 - 5 455 137
          XM_011515891.2 XP_011514193.1 2 - 3 618 137
          XM_011515892.2 XP_011514194.1 4 - 5 3002 137
          XM_011515897.2 XP_011514199.1 5 - 6 709 137
          XM_011515898.2 XP_011514200.1 5 - 6 636 137
          XM_011515899.2 XP_011514201.1 4 - 5 3004 137
          XM_011515901.2 XP_011514203.1 4 - 5 3004 137
          XM_017011817.1 XP_016867306.1 4 - 5 3000 137
          XM_017011818.1 XP_016867307.1 5 - 6 682 137
          XM_017011819.1 XP_016867308.1 4 - 5 3000 137
          XM_017011821.1 XP_016867310.1 5 - 6 709 137
          GenBank mRNA AB208895.1 - 4 - 5 492 137
          AK293239.1 - 4 - 5 469 137
          AK302216.1 - 4 - 5 658 137
          AK303585.1 - 4 - 5 496 137
          AK303637.1 - 5 - 6 710 137
          AY519465.1 - 3 - 4 365 137
          BC010858.2 - 4 - 5 487 3901250 137
          BG829016.1 - 4 - 5 532 138
          U61145.1 - 4 - 5 454 137
          X95653.1 - 4 - 5 422 137

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AK303637.1, XM_011515897.2, XM_011515898.2, XM_017011821.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Inventoried MicroArray Quality Control (MAQC) Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene EZH2

          Panther Classification:

          Molecular Function -

          histone modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          DNA methylation
          chromatin organization
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          positive regulation of epithelial to mesenchymal transition
          regulation of gliogenesis
          skeletal muscle satellite cell maintenance involved in skeletal muscle regeneration
          cerebellar cortex development
          hippocampus development
          response to estradiol
          negative regulation of transcription elongation from RNA polymerase II promoter
          regulation of cell proliferation
          regulation of circadian rhythm
          positive regulation of MAP kinase activity
          negative regulation of sequence-specific DNA binding transcription factor activity
          positive regulation of GTPase activity
          negative regulation of epidermal cell differentiation
          negative regulation of gene expression, epigenetic
          negative regulation of transcription, DNA-templated
          negative regulation of retinoic acid receptor signaling pathway
          rhythmic process
          negative regulation of striated muscle cell differentiation
          cellular response to hydrogen peroxide
          G1 to G0 transition
          histone H3-K27 methylation
          protein localization to chromatin
          positive regulation of protein serine/threonine kinase activity
          positive regulation of dendrite development
          negative regulation of G1/S transition of mitotic cell cycle
          core promoter binding
          DNA binding
          chromatin binding
          RNA binding
          protein binding
          protein-lysine N-methyltransferase activity
          histone-lysine N-methyltransferase activity
          chromatin DNA binding
          histone methyltransferase activity
          ribonucleoprotein complex binding
          sequence-specific DNA binding
          histone methyltransferase activity (H3-K27 specific)
          promoter-specific chromatin binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline