Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs00243216_s1
          See other SRY GE Assays ›
          Gene Symbol:
          SRY XGY2
          Gene Name:
          sex determining region Y
          XG Y-linked 2 (pseudogene)
          Gene Aliases:
          SRXX1, SRXY1, TDF, TDY
          XG, XGPY, XGPY2
          Chromosome Location:
          Chr.Y: 2786855 - 2787682 on Build GRCh38
          Chromosome Location:
          Chr.Y: 2752296 - 2774996 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Hs00243216_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SRY
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_003140.3 NP_003131.1 1 493 91
          GenBank mRNA BC074923.2 - 1 452 91
          BC074924.2 - 1 452 91
          L10101.1 - 1 510 91
          MH972534.1 - 1 414 91
          MH972535.1 - 1 414 91
          MH972536.1 - 1 424 91
          MH972537.1 - 1 424 91
          MK011510.1 - 1 414 91
          MK011511.1 - 1 424 91
          MK011512.1 - 1 424 91
          MK011513.1 - 1 424 91
          MK011514.1 - 1 424 91
          MK011516.1 - 1 424 91
          MK011517.1 - 1 424 91
          MK011518.1 - 1 424 91
          MK011519.1 - 1 424 91
          S53156.1 - 1 346 91
          S56543.1 - 1 249 91
          Gene Symbol: XGY2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA Z48512.1 - 4 302 91
          Z48513.1 - 4 302 91

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0Y00006909.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC0Y000121.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Human Androgen Panel Inventoried Human_Androgen Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SRY
          Click here to find siRNAs for Gene XGY2

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription from RNA polymerase II promoter
          sex differentiation
          positive regulation of gene expression
          cell differentiation
          male sex determination
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of male gonad development
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          protein binding
          calmodulin binding
          DNA-binding transcription factor binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0Y00006909.hg.1 ENST00000383070.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0Y00006909.hg.1 ENST00000383070.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0Y00006909.hg.1 ENST00000383070.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0Y00006909.hg.1 ENST00000383070.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0Y00006909.hg.1 NM_003140.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0Y00006909.hg.1 NM_003140.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0Y00006909.hg.1 NM_003140.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0Y00006909.hg.1 NM_003140.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0Y00006909.hg.1 NM_003140.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR0Y001377.hg.1 ENST00000383070.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0Y001377.hg.1 ENST00000525526.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0Y001377.hg.1 ENST00000534739.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0Y001377.hg.1 NM_003140.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR0Y001377.hg.1 TC0Y000121.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR0Y001378.hg.1 ENST00000383070.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0Y001378.hg.1 NM_003140.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR0Y001378.hg.1 TC0Y000121.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR0Y001380.hg.1 NM_003140.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR0Y001380.hg.1 TC0Y000121.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR0Y001379.hg.1 ENST00000383070.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR0Y001379.hg.1 NM_003140.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR0Y001379.hg.1 TC0Y000121.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0Y00146955.hg.1 ENST00000383070.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0Y00146955.hg.1 NM_003140.2 Refseq
          Affymetrix Human Clariom D Assay PSR0Y00146955.hg.1 ENST00000383070.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0Y00146955.hg.1 NM_003140.2 Refseq
          Affymetrix Human Clariom D Assay PSR0Y00146955.hg.1 TC0Y00006909.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0Y00146956.hg.1 ENST00000383070.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0Y00146956.hg.1 NM_003140.2 Refseq
          Affymetrix Human Clariom D Assay PSR0Y00146956.hg.1 ENST00000383070.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0Y00146956.hg.1 NM_003140.2 Refseq
          Affymetrix Human Clariom D Assay PSR0Y00146956.hg.1 TC0Y00006909.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0Y00146957.hg.1 ENST00000383070.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0Y00146957.hg.1 NM_003140.2 Refseq
          Affymetrix Human Clariom D Assay PSR0Y00146957.hg.1 ENST00000383070.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0Y00146957.hg.1 NM_003140.2 Refseq
          Affymetrix Human Clariom D Assay PSR0Y00146957.hg.1 TC0Y00006909.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0Y00146958.hg.1 NM_003140.2 Refseq
          Affymetrix Human Clariom D Assay PSR0Y00146958.hg.1 NM_003140.2 Refseq
          Affymetrix Human Clariom D Assay PSR0Y00146958.hg.1 TC0Y00006909.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0Y00006909.hg.1 ENST00000383070.1 Ensembl
          Affymetrix Human Clariom D Assay TC0Y00006909.hg.1 ENST00000383070.1 Ensembl
          Affymetrix Human Clariom D Assay TC0Y00006909.hg.1 ENST00000383070.1 Ensembl
          Affymetrix Human Clariom D Assay TC0Y00006909.hg.1 ENST00000383070.1 Ensembl
          Affymetrix Human Clariom D Assay TC0Y00006909.hg.1 NM_003140.2 Refseq
          Affymetrix Human Clariom D Assay TC0Y00006909.hg.1 NM_003140.2 Refseq
          Affymetrix Human Clariom D Assay TC0Y00006909.hg.1 NM_003140.2 Refseq
          Affymetrix Human Clariom D Assay TC0Y00006909.hg.1 NM_003140.2 Refseq
          Affymetrix Human Clariom D Assay TC0Y00006909.hg.1 NM_003140.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline